Skip to Content
Merck
All Photos(1)

Key Documents

4.90028

Sigma-Aldrich

1-Butyl-1-methylpyrrolidinium chloride

for synthesis

Synonym(s):

1-Butyl-1-methylpyrrolidinium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H20ClN
CAS Number:
Molecular Weight:
177.71
MDL number:
UNSPSC Code:
12352107

Quality Level

Assay

≥98.0% (electrophoresis)

form

solid

pH

8 (25 °C in H2O)

storage temp.

2-30°C

InChI

1S/C9H20N.ClH/c1-3-4-7-10(2)8-5-6-9-10;/h3-9H2,1-2H3;1H/q+1;/p-1

InChI key

BOOXKGZZTBKJFE-UHFFFAOYSA-M

Analysis Note

Purity (electrophoresis): ≥ 98.0 %
Identity (NMR): passes test
Water (K. F.): ≤ 1.0 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service