Skip to Content
Merck
All Photos(1)

Key Documents

1.07495

Supelco

Pyrrolidine-1-dithiocarboxylic acid ammonium salt

GR for analysis

Synonym(s):

Pyrrolidine-1-dithiocarboxylic acid ammonium salt, Ammonium-N,N-tetramethylenedithiocarbamate, N,N-Tetramethylenedithiocarbaminic acid ammoniumsalt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12N2S2
CAS Number:
Molecular Weight:
164.29
MDL number:
UNSPSC Code:
12352107
EC Index Number:
225-834-4

Quality Level

form

solid

dilution

(for analytical testing)

pH

6-7 (20 °C, 50 g/L in H2O)

mp

146-151 °C

solubility

190 g/L

bulk density

400 kg/m3

storage temp.

2-8°C

InChI

1S/C5H9NS2.H3N/c7-5(8)6-3-1-2-4-6;/h1-4H2,(H,7,8);1H3

InChI key

MDDIUTVUBYEEEM-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Identity (IR): passes test
Heavy metals (as Pb): ≤ 0.001 %
Iron (Fe): ≤ 0.001 %
Sulfated ash: ≤ 0.10 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service