Skip to Content
Merck
All Photos(1)

Key Documents

870734P

Avanti

18:0(α-OH) Coenzyme A

Avanti Research - A Croda Brand 870734P, powder

Synonym(s):

α-hydroxy stearoyl Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H79N10O18P3S
CAS Number:
Molecular Weight:
1101.09
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (870734P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870734P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(C(O)CCCCCCCCCCCCCCCC)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C39H70N7O18P3S/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-27(47)38(52)68-22-21-41-29(48)19-20-42-36(51)33(50)39(2,3)24-61-67(58,59)64-66(56,57)60-23-28-32(63-65(53,54)55)31(49)37(62-28)46-26-45-30-34(40)43-25-44-35(30)46/h25-28,31-33,37,47,49-50H,4-24H2,1-3H3,(H,41,48)(H,42,51)(H,56,57)(H,58,59)(H2,40,43,44)(H2,53,54,55)/t27-,28+,31+,32+,33-,37+/m0/s1

InChI key

OJQMIXCIJFLULT-LPSUQUFLSA-N

General description

18:0(α-OH) Coenzyme A is a qualitative standard useful for general identification of lipids by MS analysis.

Biochem/physiol Actions

CoA plays important roles in many metabolic pathways, including the tricarboxylic acid cycle, and the synthesis and oxidation of fatty acids. Acylated derivatives, for example Acetyl-CoA, are critical intermediates in many metabolic reactions ADD>, particularly fatty acid and carbohydrate metabolism.

Packaging

5 mL Amber Glass Screw Cap Vial (870734P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Continuous fatty acid oxidation and reduced fat storage in mice lacking acetyl-CoA carboxylase 2
Ab Elheiga L, et al.
Science (New York, N.Y.), 2613-2616 (2001)
Continuous fatty acid oxidation and reduced fat storage in mice lacking acetyl-CoA carboxylase 2
Ab Elheiga L, et al.
Science, 2613-2616 (2001)
Nonalcoholic fatty liver disease and mitochondrial dysfunction.
Wei Y, et al.
World Journal of Gastroenterology (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service