Skip to Content
Merck
All Photos(1)

Key Documents

300113S

Avanti

Control (2mL vial)

Avanti Research - A Croda Brand

Synonym(s):

Control (Pegylated Liposomal Vehicle), frozen liposomal suspension (buffer - 10% w/w sucrose, 10 mM histidine ~pH 6.5)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (HPLC/MS)

form

solution

packaging

pkg of 1 × 2 mL (300113S-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

advanced drug delivery

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

Formulation: A Pegylated, long-circulating liposomal formulation with an ammonium sulfate core. Liposomes are stored frozen for a longer shelf life.

Application

Control (2mL vial) may be used as a vehicle control liposome for liposomal encapsulated doxorubicin (Dox-NP).

Packaging

10 mL clear serum vial (300113S-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service