Key Documents
Z653349
Steinel® hot air guns
HL 2020 E
About This Item
UNSPSC Code:
41121800
AC/DC input
230 V
description
infinitely variable temperature
feature
CE compliant
manufacturer/tradename
HL 2020 E
availability
available only in EU
parameter
150-500 L/min flow rate
80-630 °C temperature
output
2200 watts
General description
The HL 2020 E hot air tool is extremely robust, promising to last a good 20% longer than its predecessor. Professional extras: the residual heat indicator warns the user that the delivery nozzle is still hot and can cause burns even long after the tool has been unplugged. And the joystick permits intuitive operation and use. And talking of use: the HL 2020 E hot air professional is designed for all STEINEL nozzles and, delivering hot air at a temperature of up to 630 °C, the ideal tool for shaping, desoldering printed circuit boards, welding plastics, drying filler, shrinking on cable sleeves, soldering pipes and other jobs requiring high, accurately adjustable heat as well as a powerful blower. The current working temperature and airflow rate is indicated on the LCD display which is easy to read at any light level. The tool immediately responds to heat build-up by indicating a warning triangle on the display.
Benefits
: LCD display, residual heat indicator for optimum protection, ergonomic operation by joystick, impressive 2200 watts of power, adjustable from 80 °C to 630 °C in 10 °C steps. Airflow rate adjustable in three stages (150 / 150-300 / 300-500 l/min.). With plastic case and 9 mm reduction nozzle. Optionally available fine-dust filter and HL Scan for measuring temperature at the workpiece. The mains power cord is very easy to replace by removing the cap on the handle.
Legal Information
Steinel is a registered trademark of Steinel GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)