WHA10370395
Whatman® glass microfiber filters with organic binder, Grade GF 10
roll, W × L (70 mm) (40 m), pack of 1 ea
All Photos(1)
About This Item
UNSPSC Code:
40161507
NACRES:
NB.24
Recommended Products
material
roll
packaging
pack of 1 ea
manufacturer/tradename
Whatman 10370395
W × L
(70 mm) (40 m)
General description
This filter with extreme mechanical stability and temperature resistant up to 180°C is used as a weighing aid for infrared weighing and as a roll filter in automatic air filtration units.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)