Key Documents
THS01239
ThalesNano CatCart® holder with SS connection tube
L × diam. 70 mm × 4 mm
Synonym(s):
Heterogeneous catalysis system, safe Heterogeneous catalysis
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
packaging
pack of 1 ea
manufacturer/tradename
ThalesNano THS 01239
L × diam.
70 mm × 4 mm
General description
CatCarts are suitable for use only with Thalesnano H-Cube reactors.
Legal Information
CatCart is a registered trademark of ThalesNano Nanotechnology Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)