Skip to Content
Merck
All Photos(1)

Key Documents

RNI00002

Sigma-Aldrich

di-Ad-BrettPhos-AuNCMe SbF6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H64AuF6NO2PSb
Molecular Weight:
1114.69
UNSPSC Code:
12352005
PubChem Substance ID:

form

solid

SMILES string

CC#[N+][Au].F[Sb-](F)(F)(F)(F)F.COc1ccc(OC)c(c1P(C23CC4CC(CC(C4)C2)C3)C56CC7CC(CC(C7)C5)C6)-c8c(cc(cc8C(C)C)C(C)C)C(C)C

InChI

1S/C43H61O2P.C2H3N.Au.6FH.Sb/c1-25(2)34-17-35(26(3)4)39(36(18-34)27(5)6)40-37(44-7)9-10-38(45-8)41(40)46(42-19-28-11-29(20-42)13-30(12-28)21-42)43-22-31-14-32(23-43)16-33(15-31)24-43;1-2-3;;;;;;;;/h9-10,17-18,25-33H,11-16,19-24H2,1-8H3;1H3;;6*1H;/q;;+1;;;;;;;+5/p-6

InChI key

KEKNRDOERHLNMX-UHFFFAOYSA-H

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service