Skip to Content
Merck
All Photos(1)

Key Documents

GF71405658

Brass alloy, Cu63Zn37

200mm tube, 0.61mm outside diameter, 0.18mm wall thickness, 0.25mm inside diameter, as drawn

Synonym(s):

CU027100

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
30102010
NACRES:
NA.23

form

tubes

manufacturer/tradename

Goodfellow 714-056-58

L × wall thickness

200 mm × 0.18 mm

O.D. × I.D.

0.61 mm × 0.25 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
GF71405658-10EA4061829925932
GF71405658-1EA4061825435114
GF71405658-20EA4061829976439
GF71405658-2EA4061829956240
GF71405658-5EA4061831006629

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service