GF53352884
Tantalum
foil, 8mm disks, thickness 0.015mm, 99.9%
Synonym(s):
Tantalum, TA000320, Ta
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Ta
CAS Number:
Molecular Weight:
180.95
MDL number:
UNSPSC Code:
12141741
PubChem Substance ID:
NACRES:
NA.23
vapor pressure
<0.01 mmHg ( 537.2 °C)
Assay
99.9%
form
foil
autoignition temp.
572 °F
manufacturer/tradename
Goodfellow 533-528-84
resistivity
13.5 μΩ-cm, 20°C
diam. × thickness
8 mm × 0.015 mm
bp
5425 °C (lit.)
mp
2996 °C (lit.)
density
16.69 g/cm3 (lit.)
SMILES string
[Ta]
InChI
1S/Ta
InChI key
GUVRBAGPIYLISA-UHFFFAOYSA-N
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yingxun Du et al.
Journal of hazardous materials, 225-226, 21-27 (2012-05-23)
In order to overcome the limitation of the application of nanoparticles, tantalum (oxy)nitrides nanotube arrays on a Ta foil were synthesized and introduced in vis (visible light)-Fenton-like system to enhance the degradation of atrazine. At first, the anodization of tantalum
Zhongli Cai et al.
The journal of physical chemistry. B, 109(10), 4796-4800 (2006-07-21)
Both monolayer and thick (20 microm) films of dry pGEM-3Zf(-) plasmid DNA deposited on tantalum foil were exposed to Al Kalpha X-rays (1.5 keV) for various times in an ultrahigh vacuum chamber. For monolayer DNA, the damage was induced mainly
J G Sen Gupta
Talanta, 31(12), 1053-1056 (1984-12-01)
A 40-fold increase in sensitivity obtained by using a tantalum foil lining in a pyrolytically-coated graphite furnace permitted determination of low ppm levels of cerium in most silicate rocks. A preliminary preconcentration by oxalate and hydroxide co-precipitations was used before
Zhongli Cai et al.
Radiation research, 165(3), 365-371 (2006-02-24)
Both thick and thin films of pGEMR-3Zf- plasmid DNA deposited on a tantalum foil were exposed to soft X rays (effective energy of 14.8 keV) for various times in air under a relative humidity of 45% (Gamma approximately 6, where
J G Sen Gupta
Talanta, 32(1), 1-6 (1985-01-01)
An improved graphite furnace atomic-absorption method has been developed for the determination of Sc, Y and the rare-earth elements in silicate rocks and related materials. The method, which involves the separation of the lanthanides by ion-exchange followed by their determination
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service