Skip to Content
Merck
All Photos(2)

Key Documents

GF51225814

Platinum

tube, 50mm, outside diameter 0.7mm, inside diameter 0.4mm, wall thickness 0.15mm, as drawn, 99.95%

Synonym(s):

Platinum, PT007150

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Pt
CAS Number:
Molecular Weight:
195.08
MDL number:
UNSPSC Code:
12141734
PubChem Substance ID:
NACRES:
NA.23

Assay

99.95%

form

tubes

manufacturer/tradename

Goodfellow 512-258-14

resistivity

10.6 μΩ-cm, 20°C

L × wall thickness

50 mm × 0.15 mm

O.D. × I.D.

0.7 mm × 0.4 mm

bp

3827 °C (lit.)

mp

1772 °C (lit.)

density

21.45 g/cm3 (lit.)

SMILES string

[Pt]

InChI

1S/Pt

InChI key

BASFCYQUMIYNBI-UHFFFAOYSA-N

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianbo Wu et al.
Accounts of chemical research, 46(8), 1848-1857 (2013-07-03)
An efficient oxygen reduction reaction (ORR) offers the potential for clean energy generation in low-temperature, proton-exchange membrane fuel cells running on hydrogen fuel and air. In the past several years, researchers have developed high-performance electrocatalysts for the ORR to address
Yanlong Yang et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(4), 2905-2921 (2013-12-18)
The relationship between the ERCC1/2 single nucleotide polymorphisms (SNPs) and the clinical outcomes of the platinum-based chemotherapy in the non-small cell lung cancer (NSCLC) is still inconsistent and inconclusive despite extensive investigations have been conducted to address this question. In
Yanqin Cao et al.
Journal of nanoscience and nanotechnology, 14(2), 1194-1208 (2014-04-23)
Porous platinum, that has outstanding catalytic and electrical properties and superior resistant characteristics to corrosion, has been widely applied in chemical, petrochemical, pharmaceutical, electronic, and automotive industries. As the catalytic activity and selectivity depend on the size, shape and structure
Johnson J Liu et al.
Current cancer drug targets, 12(8), 962-986 (2012-07-17)
Membrane transporters govern the movement of drugs and their metabolites across biological membranes, thereby determining their pharmacokinetics, efficacy and adverse drug reactions. Platinum-based anticancer drugs are a mainstay of chemotherapy for many human malignancies. However, their clinical utility is limited
Dalian Ding et al.
Anatomical record (Hoboken, N.J. : 2007), 295(11), 1851-1867 (2012-10-10)
Cisplatin, carboplatin, nedaplatin, and oxaliplatin are widely used in contemporary oncology; however, their ototoxic and neurotoxic side effects are quite different as discussed in this review. Cisplatin is considered the most ototoxic, but despite its reputation, the magnitude of hair

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service