GF19348892
Tungsten
foil, 0.5m coil, thickness 0.076mm, coil width 1.5mm, as rolled, 99.95%
Synonym(s):
Tungsten, W 000251, W
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
W
CAS Number:
Molecular Weight:
183.84
MDL number:
UNSPSC Code:
12141747
PubChem Substance ID:
NACRES:
NA.23
Assay
99.95%
form
foil
manufacturer/tradename
Goodfellow 193-488-92
resistivity
4.9 μΩ-cm, 20°C
L × W × thickness
0.5 m × 1.5 mm × 0.076 mm
bp
5660 °C (lit.)
mp
3410 °C (lit.)
density
19.3 g/mL at 25 °C (lit.)
SMILES string
[W]
InChI
1S/W
InChI key
WFKWXMTUELFFGS-UHFFFAOYSA-N
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Congshang Wan et al.
Ultramicroscopy, 119, 106-110 (2011-11-15)
Thermionic electron emission from 200 to 500 nm thick coatings of scandium oxide on tungsten foil have been examined in thermionic emission microscopy, spectroscopic photoelectron microcopy, synchrotron radiation and ultraviolet photoelectron spectroscopy (UPS). A clear dependence of the scandium oxide-W
J R Marbach et al.
Physics in medicine and biology, 26(3), 435-443 (1981-05-01)
Curves relating beam energy, scattering foil thickness, central-axis depth-dose, and beam flatness have been generated using data taken on a Siemens 200A betatron. The curve set allows a single combination of tungsten foil thickness and electron beam energy to be
Carlos D Brondino et al.
Current opinion in chemical biology, 10(2), 109-114 (2006-02-17)
Mononuclear molybdenum and tungsten are found in the active site of a diverse group of enzymes that, in general, catalyze oxygen atom transfer reactions. Enzymes of the xanthine oxidase family are the best-characterized mononuclear Mo-containing enzymes. Several 3D structures of
Dae Yup Han et al.
International journal of radiation oncology, biology, physics, 89(3), 666-673 (2014-04-23)
To demonstrate that utilization of the direction-modulated brachytherapy (DMBT) concept can significantly improve treatment plan quality in the setting of high-dose-rate (HDR) brachytherapy for cervical cancer. The new, MRI-compatible, tandem design has 6 peripheral holes of 1.3-mm diameter, grooved along
Gijsbert B van der Voet et al.
Military medicine, 172(9), 1002-1005 (2007-10-17)
Tungsten and tungsten compounds are considered toxicologically relatively safe. Concern regarding the potential health and environmental effects of depleted uranium and lead in military applications has lead many countries to explore the possibility of applying toxicologically safer metals. Heavy metal
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service