Skip to Content
Merck
All Photos(1)

Key Documents

DWKWGW538VTMLME

Azlon Multi-Lingual Safety Wash Bottles With Driplok Vapor Venting

label, Methanol (English, French, Arabic, Wide Neck), capacity 500 mL

Synonym(s):

WGW538VTMLME

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103209
NACRES:
NB.43

feature

label Methanol (English, French, Arabic, Wide Neck)

packaging

pkg of 5 ea

manufacturer/tradename

Azlon WGW538VTMLME

capacity

500 mL

Looking for similar products? Visit Product Comparison Guide

Application

Ideal for methanol dispensing within the laboratory

Features and Benefits

Patented Driplok vapour venting valve to prevent solvent drips. Multilingual with solvent labels including all new globally harmonised symbols, NFPA diamond, risk phrases and safety information

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service