DWKWGW234PVENT
Azlon Square Shoulder Safety Wash Bottles With Driplok Vapor Venting
wide-neck, low-density polyethylene bottle, orange polypropylene closure, capacity 500 mL
Synonym(s):
WGW234PVENT
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41103209
NACRES:
NB.43
Recommended Products
material
low-density polyethylene bottle
orange polypropylene closure
wide-neck
packaging
pkg of 5 ea
manufacturer/tradename
Azlon WGW234PVENT
capacity
500 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Ideal for solvent and water dispensing within the laboratory
Features and Benefits
Patented Driplok vapour venting valve to prevent solvent drips
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service