Skip to Content
Merck
All Photos(1)

Key Documents

CLS76662L

Pyrex® snap cap

snap cap size 2,000

Synonym(s):

polyethylene cap, polyethylene caps, stopper, Corning® 7666

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
41122500
NACRES:
NB.43

material

low-density polyethylene

snap cap size

2,000

manufacturer/tradename

Corning 7666-2L

color

blue

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service