Key Documents
BR781346
BRAND® 384-well PCR plate
new
PP, 0.03 ml, full-skirted, transparent, BIO-CERT® PCR QUALITY, 50EA
About This Item
UNSPSC Code:
41121800
material:
transparent polypropylene plate
size:
384 well
sterility:
non-sterile
binding type:
non-treated surface
feature:
DNA free
RNase and DNase free
pyrogen free
skirt
material
transparent polypropylene plate
sterility
non-sterile
feature
DNA free
RNase and DNase free
pyrogen free
skirt
packaging
pack of 50 ea
manufacturer/tradename
BRAND 781346
size
384 well
wells
384
working volume
30 μL
suitability
suitable for PCR
binding type
non-treated surface
General description
BRAND® 384-well PCR plates are an economical solution for automatic high-throughput analyses. The thin walls allow fast transmission of the temperature specified by the cycler, thus reducing cycle times. The stable design of the plates makes them ideal for automation, as they can be gripped by robot systems without twisting. To minimize evaporation with small sample volumes to 2 µl and prevent contamination, seal the plates with the appropriate sealing film. The cleanroom quality of the PCR consumables, including 384-well PCR plates from BRAND® guarantees reliable results.
Features and Benefits
  • For applications with automation and high-throughput analyses
  • Rapid heat transfer through thin vessel walls
  • Can be sealed reliably with self-adhesive sealing films
  • BIO-CERT® PCR QUALITY: Free of DNA, RNase, DNase, Pyrogens and PCR-Inhibitors
Legal Information
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)