940615
Rhodium(III) chloride

anhydrous, 48.9 ± 3.0% Rh
Synonym(s):
RhCl3, Rhodium chloride, Trichlororhodium
About This Item
Quality Level
form
solid
SMILES string
Cl[Rh](Cl)Cl
InChI
1S/3ClH.Rh/h3*1H;/q;;;+3/p-3
InChI key
SONJTKJMTWTJCT-UHFFFAOYSA-K
Looking for similar products? Visit Product Comparison Guide
General description
Application
It can be used as
- As a starting material for synthesizing 3D palladium-rhodium (PdRh) nanosheet assembly (NSA) catalysts using a seed-mediated growth method. The resulting open 3D structure significantly enhances electrocatalytic activity for the methanol oxidation reaction (MOR).
- As a precursor for the synthesis of Rhodium nanoparticles supported on phosphorus-doped carbon (Rh/PC) using the ball-milling method with sodium hypophosphite as the phosphorus source. This catalyst shows excellent conversion and high regioselectivity for the hydroformylation of styrene, outperforming phosphorus-free (Rh/C) and metal-free (PC) catalysts.
- To synthesis of heterogeneous Rh/COF (covalent organic framework) catalyst using melamine and Rhodium(III) chloride as a precursor. This catalyst enables the tandem selective transfer hydrogenation and monomethylation of nitro compounds with methanol.
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service