Skip to Content
Merck
All Photos(1)

Key Documents

940321

Sigma-Aldrich

Lithium sulfide

new

99.995% trace metals basis (basis), low sodium, battery grade

Synonym(s):

Dilithium monosulfide, Dilithium sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Li2S
CAS Number:
Molecular Weight:
45.95
MDL number:
UNSPSC Code:
12352300

grade

battery grade

Quality Level

description

application(s): battery manufacturing

Assay

99.995% trace metals basis (basis)

form

powder

mol wt

45.95

composition

Li2S

particle size

100 mesh

bp

1,372 °C ((lit.))

mp

900-975 °C

solubility

soluble (reacts with water)

density

1.66 g/mL at 25 °C (lit.)

cation traces

Na: <35 ppm

storage temp.

2-8°C

SMILES string

[Li]S[Li]

InChI

1S/2Li.S

InChI key

ZWDBUTFCWLVLCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium sulfide (Li₂S) is a high-purity, inorganic compound that appears as a white to yellowish crystalline powder. It has a molecular weight of 45.95 g/mol and a density of 1.66 g/cm³. Lithium sulfide is known for its strong nucleophilic properties and its ability to react readily with water to produce lithium hydroxide and hydrogen sulfide gas. It is also soluble in certain organic solvents, such as pyridine and dimethyl sulfoxide. Lithium sulfide crystallizes in an antifluorite structure and melts in the temperature range of 900-975°C. Our high-purity material (99.995% trace metals) is specifically formulated for low sodium content, with less than 35 ppm of sodium metal. Additionally, the material is passed through a 100 mesh sieve, ensuring that most particles are less than 150 microns in size.

Application

High-purity lithium sulfide is primarily used as a precursor in the synthesis of other lithium compounds. In chemical manufacturing, it acts as a precursor for lithium hydride, lithium borohydride, lithium amide, lithium thioacetate, lithium tetrafluoroborate, and others. In the battery industry, our battery grade lithium sulfide is used as a precursor to synthesize sulfide-based solid electrolytes like Li₃PS₄ ([https://www.sigmaaldrich.com/US/en/product/aldrich/916374]), Li₆PS₅Cl ([https://www.sigmaaldrich.com/US/en/product/aldrich/916137]), and Li₁₀SnP₂S₁₂ ([https://www.sigmaaldrich.com/US/en/product/aldrich/915114]). Lithium sulfide also serves as a cathode material in lithium-sulfur batteries, offering a theoretical capacity of up to 1166 mAh/g, nearly four times that of lithium cobalt oxide. This makes it an attractive candidate for next-generation battery technologies. Additionally, lithium sulfide is valuable in the semiconductor industry for the production of thin-film transistors (TFTs) and other electronic devices, particularly lithium sulfide-phosphorus oxide (Li₂S-P₂O₅) TFTs.

Features and Benefits

Higher Purity (99.995%): Ensures minimal trace metal contamination, making it ideal for applications requiring high-purity materials.
Low Sodium Content: Specifically formulated for low sodium content (<35 ppm) to meet the stringent requirements of sensitive applications.
Controlled Particle Size: Passed through a 100 mesh sieve, ensuring most particles are less than 150 microns, which is crucial for consistent performance in various applications.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service