Skip to Content
Merck
All Photos(1)

Key Documents

940232

Sigma-Aldrich

Ketjenblack® EC-300J

new

conductive carbon black

Synonym(s):

Carbon black

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
UNSPSC Code:
12171603

grade

battery grade

Quality Level

description

surface area: ~800 m2/g (BET)

form

powder

pH

9.0-10.5

bp

4200 °C (lit.)

mp

3550 °C (lit.)

bulk density

0.125‑0.145 g/cm3

application(s)

battery manufacturing

SMILES string

[C]

General description

Ketjenblack® EC-300J is a highly pure carbon black known for its distinctive mesoporous structure, which results in a high surface area of approximately 800 m2/g (BET) and exceptional electronic conductivity.

Application

Ketjenblack® EC-300J is a versatile carbon black suitable for a wide range of antistatic and electroconductive applications. In research and development, it serves as a valuable support for electrocatalysts in fuel cell electrodes and as a conductive additive in battery electrodes. Due to its high surface area and its capability to form electronically conductive networks, Ketjenblackket® EC-300J is an excellent support for nanoparticle catalysts. The carbon′s stability at very high temperatures (>1000°C) allows for post-synthesis thermal treatment and annealing, adding an advantage to its use. Moreover, it plays a crucial role in the fabrication of battery electrodes, especially with nanoparticle cathode active materials, enhancing the electronic conductivity of the electrode film and subsequently improving battery performance. Ketjenblack® EC-300J can be challenging to disperse, especially in viscous media. For instance, when used in battery electrodes, it is recommended to first thoroughly disperse Ketjenblack® EC-300J in the solvent before incorporating PVDF binder or the cathode active material to minimize carbon agglomeration in the final composite.

Legal Information

Ketjenblack is a registered trademark of Nouryon Functional Chemicals B.V. or affiliates

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service