939927
1,3-Propanediol cyclic sulfate

≥99.5%, ≤5 μmol/g acid, anhydrous, battery grade
Synonym(s):
1,3,2-Dioxathiane 2,2-dioxide, 1,3,2-dioxathiane-2,2-dione, PCS battery additive, Trimethylene sulfate
About This Item
grade
anhydrous
battery grade
Quality Level
Assay
≥99.5%
form
powder or crystals
impurities
≤100 ppm H2O
≤5 μmol/g acid
bp
153 °C
mp
58-62 °C (lit.)
59-62 °C
solubility
soluble (H2O: highly soluble(lit.))
soluble (carbonates: soluble(lit.))
soluble (glymes: soluble(lit.))
application(s)
battery manufacturing
SMILES string
O=S1(=O)OCCCO1
InChI
1S/C3H6O4S/c4-8(5)6-2-1-3-7-8/h1-3H2
InChI key
OQYOVYWFXHQYOP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1B
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service