MAK330
Glutamate Assay Kit
sufficient for 100 colorimetric tests
Synonym(s):
Glutamate Detection Kit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12161503
NACRES:
NA.84
Recommended Products
usage
sufficient for 100 colorimetric tests
input
plasma
serum
cell medium
application(s)
cosmetics
food and beverages
pharmaceutical
detection method
colorimetric
relevant disease(s)
neurological disorders
storage temp.
−20°C
Related Categories
General description
Glutamate is an important chemical in general metabolism. It is also a crucial mammalian neurotransmitter that is believed to be involved in a number of neurological and psychiatric disorders such as lateral sclerosis, lathyrism, autism and Alzheimer′s disease. Glutamate is also widely used as a flavor enhancer in the food industry. Simple, direct and automation-ready procedures for measuring glutamate concentration are very desirable.
Application
The Glutamate Assay Kit may be used for:
- Drug Discovery
- Food and Beverage Testing
- Neurological Disease Research
Features and Benefits
Broad Detection Range: Detect Glutamate levels accurately across a wide range, from 50 µM to 2.5 mM, using only 20 μL of sample in a 96-well plate setup, ensuring comprehensive analysis with minimal sample requirement.
Simplified Process: Experience a streamlined process with the addition of only a single working reagent and a 30 minute room temperature reaction, reducing complexity and saving valuable time and effort.
Compatibility with High-Throughput Systems: Easily incorporate our kit into high-throughput handling systems, ensuring smooth and accurate processing, enhancing efficiency in your laboratory workflow.
Simplified Process: Experience a streamlined process with the addition of only a single working reagent and a 30 minute room temperature reaction, reducing complexity and saving valuable time and effort.
Compatibility with High-Throughput Systems: Easily incorporate our kit into high-throughput handling systems, ensuring smooth and accurate processing, enhancing efficiency in your laboratory workflow.
Suitability
Suitable for glutamate determination in serum, plasma, tissue extracts and food extract samples. Also suitable in drug discovery/pharmacology for studying the effects of drugs on glutamate levels.
Principle
The Glutamate Assay Kit is based on glutamate dehydrogenase catalyzed oxidation of glutamate, in which the formed NADH reduces a formazan (MTT) Reagent. The intensity of the product color, measured at 565 nm, is proportionate to the glutamate concentration in the sample. Detection limit of 50 μM , linearity up to 2.5 mM glutamate in 96-well plate assay.
Other Notes
For additional information on our range of Biochemicals, please complete this form.
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service