HPLC Analysis of Tartaric Acid Enantiomers on Astec® CLC-D, Trace Level Detection
Materials
Analytical column
Product No.
Description
Pricing
CONDITIONS
column
Astec CLC-D, 15 cm x 4.6 mm I.D., 5 μm particles (53023AST)
mobile phase
3 mM copper sulfate, pH 3.2
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
10 μL
sample
5 mg/mL in water