Merck
HPLC Analysis of p-Toluenesulfonic Acid (PTSA) and Pyrimethamine on SeQuant® ZIC-HILIC
HPLC Analysis of p-Toluenesulfonic Acid (PTSA) and Pyrimethamine on SeQuant® ZIC-HILIC application for HPLC
CONDITIONS
column
SeQuant ZIC-HILIC, PEEK, 150 x 4.6 mm, 5 μm, 200 Å (1.50455.0001)
mobile phase
[A] acetonitrile; [B] ammonium acetate; (90:10, A:B)
flow rate
0.5 mL/min
column temp.
ambient
detector
254 nm
injection
1 μL
Description
Legal Information
SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany