MilliporeSigma
HPLC Analysis of Cresols and Phenol on Astec® CYCLOBOND I 2000
HPLC Analysis of Cresols and Phenol on Astec® CYCLOBOND I 2000 application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
Astec® CYCLOBOND I 2000 Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
CONDITIONS
column
CYCLOBOND I 2000, 25 cm x 4.6 mm I.D., 5 μm particles (20024AST)
mobile phase
[A] water; [B] methanol; (90:10, A:B)
flow rate
1 mL/min
column temp.
22 °C
detector
UV, 254 nm
injection
1 μL
sample
0.5 mg/mL in methanol
peak 1: o-cresol (10.70 min)
peak 2: phenol (11.70 min)
peak 3: m-cresol (14.20 min)
peak 4: p-cresol (20.10 min)
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
null