HPLC Analysis of Benzalkonium Chloride in Commercial Disinfecting Wipes on Ascentis® Express HILIC
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Ascentis Express HILIC, 15 cm x 4.6 mm I.D., 2.7 μm particles (53981-U)
mobile phase
[A] 20 mM ammonium acetate; [B] acetonitrile; (10:90, A:B)
flow rate
1 mL/min
column temp.
35 °C
detector
ESI(+), TIC 150 - 500 m/z
injection
2 μL
sample
Single wipe sheet, extracted with 10 mL methanol
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany