HPLC Analysis of Azithromycin on Ascentis® Express C18 5μm
Materials
analytical column
Product No.
Description
Pricing
mobile phase component
Product No.
Description
Pricing
standard
Product No.
Description
Pricing
CONDITIONS
column
Ascentis Express C18, 15 cm x 4.6 mm I.D., 5 μm particles (50537-U)
mobile phase
[A] 10 mM potassium phosphate, dibasic, pH 7.0 with phosphoric acid; [B] methanol; (20:80, A:B)
flow rate
1.5 mL/min
pressure
2558 psi (176 bar)
column temp.
45 °C
detector
UV, 210 nm
injection
10 μL
Description
Analysis Note
A simple high-performance liquid chromatography (HPLC) method was developed for the determination of azithromycin in raw materials and pharmaceutical formulations without any pretreatment or derivatization step.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany