HPLC Analysis of Aniline Homologs on Discovery® C18
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery C18, 15 cm × 4.6 mm I.D., 5 μm particles (504955)
mobile phase
[A] methanol; [B] water (60:40, A:B)
flow rate
1 mL/min
column temp.
30 °C
detector
UV, 254 nm
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany