MilliporeSigma
HPLC Analysis of HPLC Protein Standard on BIOshell A400 Protein C4
HPLC Analysis of HPLC Protein Standard on BIOshell A400 Protein C4 application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
BIOshell A400 Protein C18 Column
L × I.D. 10 cm × 2.1 mm, 3.4 μm particle size
standard
Product No.
Description
Pricing
HPLC protein standard mixture
analytical standard
CONDITIONS
column
BIOShell A400 C18, 10 cm x 2.1 mm I.D., 3.4 μm particles (67463-U)
mobile phase
[A] (water, 0.1% TFA); (acetonitrile, 0.08% TFA), 75:25; [B] (water, 0.1% TFA); (acetonitrile, 0.08% TFA), 25:75
flow rate
0.3 mL/min
gradient
0 to 75% B in 15 min
pressure
2800 psi (193 bar)
column temp.
30 °C
detector
UV, 215 nm
injection
2 μL
sample
ribonuclease A, cytochrome c, holo-transferrin and myoglobin 1 mg in mobile phase A
Description
Analysis Note
Analysis of protein standard of various hydrophobicity on BIOshell A400 Protein C18 to compare the selectivity and retention times.
Legal Information
null