GC Analysis of Sugars (as Alditol Acetates) on Equity-1701
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Equity-1701, 30 m × 0.25 mm I.D., 0.25 μm (28372-U)
oven
220 °C
inj. temp.
220 °C
detector
FID, 260 °C
carrier gas
helium, 20 cm/sec @ 220 °C
injection
1 μL 25:1 split
sample
Alditol acetate mixes @ 500 ppm in methylene chloride (47880-U & 47881)
Description
Legal Information
null