GC Analysis of FAMEs in Five Edible Oils (Canola, Lard, Palm, Soybean, and Sunflower Seed) on SLB®-IL111
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SLB-IL111, 30 m x 0.25 mm I.D., 0.20 μm (28927-U)
oven
180 °C
inj. temp.
250 °C
detector
FID, 260 °C
carrier gas
helium, 25 cm/sec
injection
1 μL, 50:1 split
liner
4 mm I.D., split type, cup design
sample
Canola (46961), Lard (47115-U), Palm (46962), Soybean (47122), and Sunflower Seed (47123) characterized reference oils, methylated using BF3-methanol prior to analysis
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany