GC Analysis of Anilines on Equity-5
Materials
Analytical column
Product No.
Description
Pricing
CONDITIONS
column
Equity-5, 30 m × 0.25 mm I.D., 0.25 μm (28089-U)
oven
50 °C (2 min), 10 ºC/min to 200 °C, 15 ºC/min to 325 °C
inj. temp.
250 °C
detector
FID, 325 °C
carrier gas
helium, constant flow, 1.3 mL/sec
injection
1 μL, splitless (0.5 min)
liner
4 mm I.D., single taper
sample
50 ng on-column of a custom anilines mix
Description
Legal Information
null