MilliporeSigma
HomeGas Chromatography (GC) GC Analysis of a 37-Component FAME Mix on Omegawax® (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of a 37-Component FAME Mix on Omegawax® (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of a 37-Component FAME Mix on Omegawax® (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
Description
Analyte
  • cis-11,14,17-Eicosatrienoic acid methyl ester
  • cis-4,7,10,13,16,19-Docosahexaenoic acid methyl ester
  • cis-8,11,14-Eicosatrienoic acid methyl ester
  • Methyl all-cis-5,8,11,14,17-eicosapentaenoate
  • Methyl arachidate
  • Methyl arachidonate
  • Methyl behenate
  • Methyl butyrate
  • Methyl caprylate
  • Methyl decanoate
  • Methyl elaidate
  • Methyl erucate
  • Methyl heneicosanoate
  • Methyl heptadecanoate
  • Methyl hexanoate
  • Methyl lactate
  • Methyl linoleate
  • Methyl linolelaidate
  • Methyl linolenate
  • Methyl myristate
  • Methyl myristoleate
  • Methyl oleate
  • Methyl palmitate
  • Methyl palmitoleate
  • Methyl pentadecanoate
  • Methyl stearate
  • Methyl tetracosanoate
  • Methyl tricosanoate
  • Methyl tridecanoate
  • Methyl undecanoate
  • Methyl γ-linolenate
  • Methyl cis,cis-11,14-eicosadienoate
  • Methyl cis-10-heptadecenoate
  • Methyl cis-10-pentadecenoate
  • Methyl cis-11-eicosenoate
  • Methyl cis-13,16-docosadienoate
  • Methyl cis-15-tetracosenoate