MilliporeSigma
MSMLS
Sigma-Aldrich
Mass Spectrometry Metabolite Library
Supplied by IROA Technologies
Synonym(s):
MSMLS, Mass Spectrometry Metabolite Libarary of Standards, Metabolic Library, Metabolite Library
Sign Into View Organizational & Contract Pricing
About This Item
UNSPSC Code:
41116107
NACRES:
NA.25
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
packaging
pkg of 7 plates
manufacturer/tradename
IROA Technologies
plate size
96 wells
suitability
suitable for mass spectrometry
application(s)
lipidomics
metabolomics
microbiology
shipped in
dry ice
storage temp.
−20°C
General description
MSMLS compounds are conveniently provided at 5μg per well, enough for multiple injections. These are high purity (>95%) compounds supplied in an economical, ready to use format. Compounds are arranged in 7 polypropylene racks. Metabolite descriptors provided with product include: Name, Parent CID, molecular formula, molecular weight, CAS, ChEBI, HMDB ID, PubChem Compound and Substance ID.
MSMLS (Mass Spectrometry Metabolite Library) is a collection of high quality small biochemical molecules used for mass spectrometry metabolomics applications and provides a broad representation of primary metabolism.
Application
Mass Spectrometry Metabolite Library is most commonly used to provide retention times and spectra for key metabolic compounds, help optimize analytical mass spectrometry protocols and qualify and quantify mass spectrometry sensitivity and limit of detection.
Features and Benefits
Over 600 unique small molecule metabolites organized in a 96-well format. To view a complete list of compounds in the library for recent lots, download the plate maps below:
  • MSMLS-LOT-230-23
  • MSMLS-LOT-230-16
  • MSMLS-LOT-230-13
    The complete plate map with Name, parent CID, molecular formula, molecular weight, CAS, ChEBI, HMDB ID, PubChem Compound and Substance ID are provided with the software download.
    Broad metabolite spectrum, key primary metabolites and intermediates covering key metabolic pathways, including the following classes of compounds:
  • Carboxylic acids, amino acids
  • Biogenic amines, polyamines
  • Nucleotides, coenzymes and vitamins
  • Mono- and disaccharides
  • Fatty acids, lipids, steroids and hormones
Suitability
Suitable for manual and automated work flows.
Preparation Note
MSMLS compounds are conveniently provided at 5μg per well, enough for multiple injections. Compounds can either be used as standards and injected individually or a strategy may be employed to pool compounds by row to allow multiple mixtures to be analyzed per injection. For more information on MSMLS, solubilization and preparation notes please refer to the product information sheet at the top of the page.
Other Notes
For additional information on our range of Biochemicals, please complete this form.
To download the software package, click here - MLSDiscovery software.
To view the MLSDiscovery Video visit: MLSDiscovery Tutorial Video.
Recommended products
A new metabolite library kit that will feature over 500 unique small molecule metabolites, containing 1 mg of each metabolite. This library of standards is most commonly used for metabolite qualification and quantification for MS and NMR, functional cellular assays and phenotypic screening. Click here to get more information.
Legal Information
MSMLS is a trademark of IROA Technologies LLC
Still not finding the right product?
signalword
Danger
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Lact. - Muta. 2 - Repr. 1A - Resp. Sens. 1 - Skin Corr. 1 - Skin Sens. 1 - STOT RE 1 - STOT RE 2 - STOT RE 2 Oral - STOT SE 2 - STOT SE 3
target_organs
Blood, Immune system, Respiratory system, Thyroid,Cardio-vascular system,Kidney
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
flash_point_f
-40.0 °F
flash_point_c
-40 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library