Enter the New Era of Media Preparation
Dispense up to 100 L of preheated, sterile & ISO compliant culture media with this bag and our new ReadyStream® System!
ON-DEMAND CULTURE MEDIA PREPARATION
The revolutionary new ReadyStream® system prepares and dispenses up to 100 liters of pre-heated culture media on demand, when and where you need it, at the push of a button.
We came up with this true innovation to save you time, energy, space, and resources after assessing the pains you have with current media preparation procedures.
With the ReadyStream®system:
- No handling of dehydrated media
- No autoclaving
- No media kitchen needed
- No bottles, no washing
- Small footprint
- Less variability of media quality
- Media readily heated for use
- Up to 100 L of ready-to-use culture media, and nothing you can’t carry
The ReadyStream® system reinvents media preparation and dispensing, the core principle is that the ReadyStream® system dilutes concentrated media with sterile water. This has huge advantages.
The main advantages of the ReadyStream® system
10x Media Concentrate at the Ready
The ReadyStream® system offers on-demand, concentrated media that gets diluted with sterile water to the correct concentration and dispense media into your sample bag.
A 3-liter ReadyStream® media bag lets you dispense up to 30 liters of culture media and a 10-liter bag up to 100 liters. The 10x concentrated media can be maintained for up to 5 days.
No Need to Autoclave Media
To create the concentrate, the water is sterilized by the ReadyStream® filter set before entering the ReadyStream® media bag containing the irradiated and ISO-compliant GranuCult® dehydrated culture media. After this step, which takes 20 minutes, the concentrate will allow you to prepare up to 100 liters of media on demand.
Instant Preheated Culture Media
Any time you need the ReadyStream® system dispenses pre-heated culture media at a preset temperature of between 30 and 42 °C and a volume of between 90 to 9999 mL.
Convenient Integration Into Your Existing Lab Setup
Connect the ReadyStream® system to any dilutor through an intermediate bottle serving as media reservoir. This allows multiple setups for greater flexibility, e.g. transportation of the bottle within the lab and connection to multiple dilutors. Also, it can facilitate the addition of supplements.
Watch the Webinar on the ReadyStream® System
Watch our on-demand webinar on ReadyStream® system, which showcases the live demo of the system and how to use it. Also, see our experts answer some of the questions raised in the session. However, you can still share your questions just register to our platform to get access to the webinar and simply submit your questions.
Related Products
Related Resources
- Brochure: ReadyStream® system
The ReadyStream® system - enter the new era of media preparation.
- Comparative Study: ReadyStream® Footprint Analysis vs. Others
A comparative study on time, energy and water requirements, performed by an independent food QC laboratory.
- Datasheet: ReadyStream® Filter Set
The ReadyStream® filter set is a sterilizing filter for deionized water to be used together with ReadyStream® media bags on the ReadyStream® system.
- Flyer: ReadyStream® & Milli-Q® Systems
The seamless connection between ReadyStream® and Milli-Q® systems can support your lab to integrate preparation workflows and maximize space.
- Quick Start Guide: ReadyStream® System
Dispensing Media and Filling Sample Bags on the ReadyStream® System
- Datasheet: ReadyStream® Media Bag GranuCult® BPW
For the preliminary non-selective enrichment of bacteria, particularly pathogenic Enterobacteriaceae such as Salmonella and Cronobacter, from food and animal feed, water and other materials.
- Datasheet: ReadyStream® Media Bag GranuCult® TSB
For the preliminary non-selective enrichment of bacteria, especially Salmonella, from food and other materials.
- Datasheet: ReadyStream® Data Matrix Generator Tool
- On-demand Webinar: ReadyStream® System
During this webinar, you will understand why we are saying that with our ReadyStream® system you will enter into a new era of media preparation. Ready to change the way you are making media?
- Application Note EN ISO 11133
See how two water sources compare when preparing and performance testing microbiological culture media acc. to EN ISO 11133: centrally purified water vs. water from a Milli-Q® IX system were assessed. Tests included media specific for Listeria and Salmonella pathogens.