MilliporeSigma
HomeGel ElectrophoresisRunning a Multiphor II Protocol
Running a Multiphor II Protocol
Ensure that the electrodes in the Immobiline DryStrip tray are connected and place the lid on the Multiphor II unit. Connect the leads on the lid to the power supply. Ensure that the current check on the EPS 3501 XL Power Supply is switched off. Begin IEF.
As isoelectric focusing proceeds, the bromophenol blue tracking dye migrates toward the anode. Note that the dye front leaves the Immobiline DryStrip gel well before focusing is complete, so clearing of the dye is no indication that the sample is focused. If the dye does not migrate, no current is flowing. If this occurs, check the contact between the electrodes and the electrode strips. Also check that the electrode leads and bridging cable are correctly connected, and check that the electrodes are positioned properly so that the marked side contacts the side rail.
The protocols below are suitable for running 7–24-cm Immobiline DryStrip gels on the Multiphor II Electrophoresis System connected to EPS 3501 XL Power Supply.
The focusing times in the tables below are guidelines only. They may vary with the nature of the sample and how the sample is applied. If using crude samples with high protein and salt content or using paper-bridge loading, the run time in total kilovolt-hours should be increased by 10%.
Guidelines for running Immobiline DryStrip gels on Multiphor II Electrophoresis System. Running conditions: Temperature 20 °C; current 2 mA total; power 5 W total. Program EPS 3501 XL Power Supply in gradient mode and with current check option turned off.