MilliporeSigma
HPLC Analysis of Benzene and Deuterated Benzene on Ascentis® Express C18
HPLC Analysis of Benzene and Deuterated Benzene on Ascentis® Express C18 application for HPLC
Materials
Analytical column
Product No.
Description
Pricing
Ascentis® Express C18, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Ascentis Express C18, 15 cm × 4.6 mm I.D., 2.7 μm particles (53829-U)
mobile phase
[A] acetonitrile; [B] water (55:45, A:B)
flow rate
1.0 mL/min
column temp.
50 °C
detector
UV, 254 nm
injection
10 μL
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany