MilliporeSigma
US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms, Comparison to Three Competitive Columns
US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms, Comparison to Three Competitive Columns suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-5ms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
SLB-5ms 30 m x 0.25 mm I.D., 0.25 μm (28471-U)
oven
40 °C (3 min), 20 °C/min to 100 °C, 10 °C/min to 200 °C, 30 °C/min to 325 °C (5 min)
inj. temp.
250 °C
MSD interface
325 °C
scan range
40-450 amu
carrier gas
helium, pressure programmed, 20 psi (0 min), 99 psi/min to 80 psi (0 min), 99 psi/min to 16.5 psi (3 min), 99 psi/min to 25 psi (hold remainder of run)
injection
1.0 μL splitless (0.75 min)
liner
4 mm I.D., single taper
sample
5 ng on-column of a 74 component semivolatile standard and 8 surrogate compounds, plus 6 internal standards (at 50 ng on-column)
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany