MilliporeSigma
GC Analysis of Scotch Spearmint Essential Oil on SUPELCOWAX 10
GC Analysis of Scotch Spearmint Essential Oil on SUPELCOWAX 10 suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SUPELCOWAX 10 Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
CONDITIONS
column
SUPELCOWAX 10, 30 m x 0.25 mm I.D., 0.25 μm (24079)
oven
75 °C (4 min), then 4 °C/min to 200 °C (10 min)
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 25cm/ sec set at 110°C
injection
0.1 μL 100:1 split
liner
4 mm I.D. cup injector
sample
Scotch Spearmint Oil, (A.M. Todd Sample), neat
Description
Legal Information
null