MilliporeSigma
GC Analysis of D-Carvone Enantiomer in Dill Seed Essential Oil on Astec® CHIRALDEX G-TA
GC Analysis of D-Carvone Enantiomer in Dill Seed Essential Oil on Astec® CHIRALDEX™ G-TA suitable for GC
Materials
Analytical column
Product No.
Description
Pricing
Astec® CHIRALDEX G-TA Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.12 μm
CONDITIONS
column
CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 μm (73033AST)
oven
40 °C (1 min), then 2 °C/min to 170 °C (15 min)
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 25cm/ sec set at 110°C
injection
0.1 μL 100:1 split
liner
4 mm I.D. cup injector
sample
Dill Seed Oil, neat
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC