MilliporeSigma
GC Analysis of Amphetamines in Plasma on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm) after SPME using 100 μm PDMS Fiber, Fast GC Analysis
GC Analysis of Amphetamines in Plasma on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm) after SPME using 100 μm PDMS Fiber, Fast GC Analysis suitable for GC, application for SPME
Materials
SPME fiber
Product No.
Description
Pricing
SPME fiber assembly Polydimethylsiloxane (PDMS)
df 100 μm(PDMS, needle size 24 ga, for use with manual holder
CONDITIONS
SPME fiber
100 μm PDMS (57300-U)
extraction
headspace, 300 rpm stirring at 75 °C for 15 min
column
SLB-5ms, 20 m x 0.18 mm I.D. x 0.36 μm (28576-U)
oven
75 °C (1 min), 10 °C/min to 190°C, 25 °C/min to 300 °C (4 min)
MSD interface
300 °C
carrier gas
helium, 0.7 mL/min. constant
liner
0.75 mm I.D. SPME
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany