MilliporeSigma
  • Acetonitrile extraction coupled with UHPLC-MS/MS for the accurate quantification of 17 heterocyclic aromatic amines in meat products.
Acetonitrile extraction coupled with UHPLC-MS/MS for the accurate quantification of 17 heterocyclic aromatic amines in meat products.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2017-10-27)
Yan Yan, Shuang Zhang, Guan-Jun Tao, Feng-Hui You, Jie Chen, Mao-Mao Zeng
ABSTRACT
This study proposed a simple and accurate acetonitrile extraction pretreatment method coupled with ultrahigh-performance liquid chromatography with tandem mass spectrometry for the simultaneous determination of 17 heterocyclic aromatic amines (HAAs) in meat products. With this new method, all 17 HAAs, including 11 polar and 6 nonpolarHAAs, were simultaneously extracted by acetonitrile and purified by one-step Oasis MCX cartridge purification. Compared with two different improved reference methods, the acetonitrile method could obtain higher recoveries (in the range of 42.5% to 99.0%) and better repeatability (lower than 12.2%). The limits of quantification were calculated between 0.028ngg
Sigma-Aldrich
4-Azabenzimidazole, 99%
Sigma-Aldrich
2-Amino-5-phenylpyridine, AldrichCPR
Sigma-Aldrich
2-Phenylpyridine, 98%