MilliporeSigma
C9orf72 expansion disrupts ATM-mediated chromosomal break repair.
Nature neuroscience (2017-07-18)
Callum Walker, Saul Herranz-Martin, Evangelia Karyka, Chunyan Liao, Katherine Lewis, Waheba Elsayed, Vera Lukashchuk, Shih-Chieh Chiang, Swagat Ray, Padraig J Mulcahy, Mateusz Jurga, Ioannis Tsagakis, Tommaso Iannitti, Jayanth Chandran, Ian Coldicott, Kurt J De Vos, Mohamed K Hassan, Adrian Higginbottom, Pamela J Shaw, Guillaume M Hautbergue, Mimoun Azzouz, Sherif F El-Khamisy
ABSTRACT
Hexanucleotide repeat expansions represent the most common genetic cause of amyotrophic lateral sclerosis (ALS) and frontotemporal dementia, though the mechanisms by which such expansions cause neurodegeneration are poorly understood. We report elevated levels of DNA-RNA hybrids (R-loops) and double strand breaks in rat neurons, human cells and C9orf72 ALS patient spinal cord tissues. Accumulation of endogenous DNA damage is concomitant with defective ATM-mediated DNA repair signaling and accumulation of protein-linked DNA breaks. We reveal that defective ATM-mediated DNA repair is a consequence of P62 accumulation, which impairs H2A ubiquitylation and perturbs ATM signaling. Virus-mediated expression of C9orf72-related RNA and dipeptide repeats in the mouse central nervous system increases double strand breaks and ATM defects and triggers neurodegeneration. These findings identify R-loops, double strand breaks and defective ATM-mediated repair as pathological consequences of C9orf72 expansions and suggest that C9orf72-linked neurodegeneration is driven at least partly by genomic instability.
Sigma-Aldrich
Anti-ubiquityl-Histone H2A Antibody, clone E6C5, clone E6C5, Upstate®, from mouse
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-GAPDH Mouse mAb (6C5), liquid, clone 6C5, Calbiochem®
Sigma-Aldrich
Anti-NBS1 (Nibrin) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution