MilliporeSigma
  • Quantification and Characterization of UVB-Induced Mitochondrial Fragmentation in Normal Primary Human Keratinocytes.
Quantification and Characterization of UVB-Induced Mitochondrial Fragmentation in Normal Primary Human Keratinocytes.
Scientific reports (2016-10-13)
Romain Jugé, Josselin Breugnot, Célia Da Silva, Sylvie Bordes, Brigitte Closs, Abdel Aouacheria
ABSTRACT
UV irradiation is a major environmental factor causing skin dryness, aging and cancer. UVB in particular triggers cumulative DNA damage, oxidative stress and mitochondrial dysfunction. The objective of our study was to provide both qualitative and quantitative analysis of how mitochondria respond to UVB irradiation in normal human epidermal keratinocytes (NHEK) of healthy donors, with the rationale that monitoring mitochondrial shape will give an indication of cell population fitness and enable the screening of bioactive agents with UVB-protective properties. Our results show that NHEK undergo dose-dependent mitochondrial fragmentation after exposure to UVB. In order to obtain a quantitative measure of this phenomenon, we implemented a novel tool for automated quantification of mitochondrial morphology in live cells based on confocal microscopy and computational calculations of mitochondrial shape descriptors. This method was used to substantiate the effects on mitochondrial morphology of UVB irradiation and of knocking-down the mitochondrial fission-mediating GTPase Dynamin-related protein 1 (DRP1). Our data further indicate that all the major mitochondrial dynamic proteins are expressed in NHEK but that their level changes were stronger after mitochondrial uncoupler treatment than following UVB irradiation or DRP1 knock-down. Our system and procedures might be of interest for the identification of cosmetic or dermatologic UVB-protective agents.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-Involucrin antibody produced in mouse, clone SY5, ascites fluid