MilliporeSigma
  • Caspase-3/-7-Specific Metabolic Precursor for Bioorthogonal Tracking of Tumor Apoptosis.
Caspase-3/-7-Specific Metabolic Precursor for Bioorthogonal Tracking of Tumor Apoptosis.
Scientific reports (2017-12-02)
Man Kyu Shim, Hong Yeol Yoon, Sangmin Lee, Mun Kyeong Jo, Jooho Park, Jong-Ho Kim, Seo Young Jeong, Ick Chan Kwon, Kwangmeyung Kim
ABSTRACT
Apoptosis is one of the most important intracellular events in living cell, which is a programmed cell death interrelated with caspase enzyme activity for maintaining homeostasis in multicellular organisms. Therefore, direct apoptosis imaging of living cells can provide enormous advantages for diagnosis, drug discovery, and therapeutic monitoring in various diseases. However, a method of direct apoptosis imaging has not been fully validated, especially for live cells in in vitro and in vivo. Herein, we developed a new apoptosis imaging technology via a direct visualization of active caspase-3/-7 activity in living cells. For this, we synthesized a caspase-3/-7-specific cleavable peptide (KGDEVD) conjugated triacetylated N-azidoacetyl-D-mannosamine (Apo-S-Ac
Sigma-Aldrich
4-Aminobenzyl alcohol, 98%
Sigma-Aldrich
2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, ≥99%