MilliporeSigma
  • SQSTM1/p62-mediated autophagy compensates for loss of proteasome polyubiquitin recruiting capacity.
SQSTM1/p62-mediated autophagy compensates for loss of proteasome polyubiquitin recruiting capacity.
Autophagy (2017-08-10)
Alik Demishtein, Milana Fraiberg, Dikla Berko, Boaz Tirosh, Zvulun Elazar, Ami Navon
ABSTRACT
Protein homeostasis in eukaryotic cells is regulated by 2 highly conserved degradative pathways, the ubiquitin-proteasome system (UPS) and macroautophagy/autophagy. Recent studies revealed a coordinated and complementary crosstalk between these systems that becomes critical under proteostatic stress. Under physiological conditions, however, the molecular crosstalk between these 2 pathways is still far from clear. Here we describe a cellular model of proteasomal substrate accumulation due to the combined knockdown of PSMD4/S5a and ADRM1, the 2 proteasomal ubiquitin receptors. This model reveals a compensatory autophagic pathway, mediated by a SQSTM1/p62-dependent clearance of accumulated polyubiquitinated proteins. In addition to mediating the sequestration of ubiquitinated cargos into phagophores, the precursors to autophagosomes, SQSTM1 is also important for polyubiquitinated aggregate formation upon proteasomal inhibition. Finally, we demonstrate that the concomitant stabilization of steady-state levels of ATF4, a rapidly degraded transcription factor, mediates SQSTM1 upregulation. These findings provide new insight into the molecular mechanisms by which selective autophagy is regulated in response to proteasomal overflow.
Sigma-Aldrich
IGEPAL® CA-630
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
TWEEN® 20, viscous liquid
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Tunicamycin Ready Made Solution, 5 mg/mL in DMSO, from Streptomyces sp.
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid