MilliporeSigma
  • Replication Stress Leading to Apoptosis within the S-phase Contributes to Synergism between Vorinostat and AZD1775 in HNSCC Harboring High-Risk TP53 Mutation.
Replication Stress Leading to Apoptosis within the S-phase Contributes to Synergism between Vorinostat and AZD1775 in HNSCC Harboring High-Risk TP53 Mutation.
Clinical cancer research : an official journal of the American Association for Cancer Research (2017-08-10)
Noriaki Tanaka, Ameeta A Patel, Lin Tang, Natalie L Silver, Antje Lindemann, Hideaki Takahashi, Roman Jaksik, Xiayu Rao, Nene N Kalu, Tseng-Cheng Chen, Jiping Wang, Mitchell J Frederick, Faye Johnson, Frederico O Gleber-Netto, Siqing Fu, Marek Kimmel, Jing Wang, Walter N Hittelman, Curtis R Pickering, Jeffrey N Myers, Abdullah A Osman
ABSTRACT
Purpose: The cure rate for patients with advanced head and neck squamous cell carcinoma (HNSCC) remains poor due to resistance to standard therapy primarily consisting of chemoradiation. As mutation of TP53 in HNSCC occurs in 60% to 80% of non-HPV-associated cases and is in turn associated with resistance to these treatments, more effective therapies are needed. In this study, we evaluated the efficacy of a regimen combining vorinostat and AZD1775 in HNSCC cells with a variety of p53 mutations.Experimental Design: Clonogenic survival assays and an orthotopic mouse model of oral cancer were used to examine the in vitro and in vivo sensitivity of high-risk mutant p53 HNSCC cell lines to vorinostat in combination with AZD1775. Cell cycle, replication stress, homologous recombination (HR), live cell imaging, RNA sequencing, and apoptosis analyses were performed to dissect molecular mechanisms.Results: We found that vorinostat synergizes with AZD1775 in vitro to inhibit growth of HNSCC cells harboring high-risk mutp53. These drugs interact synergistically to induce DNA damage, replication stress associated with impaired Rad51-mediated HR through activation of CDK1, and inhibition of Chk1 phosphorylation, culminating in an early apoptotic cell death during the S-phase of the cell cycle. The combination of vorinostat and AZD1775 inhibits tumor growth and angiogenesis in vivo in an orthotopic mouse model of oral cancer and prolongs animal survival.Conclusions: Vorinostat synergizes with AZD1775 in HNSCC cells with mutant p53 in vitro and in vivo A strategy combining HDAC and WEE1 inhibition deserves further clinical investigation in patients with advanced HNSCC. Clin Cancer Res; 23(21); 6541-54. ©2017 AACR.
Sigma-Aldrich
SAHA, ≥98% (HPLC)
Sigma-Aldrich
Anti-p21WAF1 (Ab-1) Mouse mAb (EA10), liquid, clone EA10, Calbiochem®
Sigma-Aldrich
Anti-Replication Protein A Antibody, clone RPA34-20, clone RPA34-20, from mouse
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid