MilliporeSigma
DJ-1 activates autophagy in the repression of cardiac hypertrophy.
Archives of biochemistry and biophysics (2017-09-25)
Ruicong Xue, Jingzhou Jiang, Bin Dong, Weiping Tan, Yu Sun, Jingjing Zhao, Yili Chen, Yugang Dong, Chen Liu
ABSTRACT
Cardiac hypertrophy is the risk factor of heart failure when the heart is confronted with pressure overload or neurohumoral stimuli. Autophagy, a conserved degradative pathway, is one of the important mechanisms involved in the regulation of cardiac hypertrophy. DJ-1 is a traditional anti-oxidative protein and emerging evidence suggested that DJ-1 might modulate autophagy. However, the regulation of autophagy by DJ-1 in the process of cardiac hypertrophy remains unknown. In our study, we firstly discovered that the expression of DJ-1declined in the process of pressure overload cardiac hypertrophy, and its alteration was parallel with the impairment of autophagy. Furthermore, we proved that DJ-1 knockout mice exhibited a more hypertrophied phenotype than wildtype mice in cardiac hypertrophy which indicated that DJ-1 is responsible for the repression of cardiac hypertrophy. Furthermore, DJ-1 knockout significantly exacerbated pulmonary edema due to cardiac hypertrophy. In the process of cardiac hypertrophy, DJ-1 knockout significantly impaired autophagy activation and enhanced mTORC1 and mTORC2 phosphorylation were found. Similarly, our in vitro study proved that DJ-1 overexpression ameliorated phenylephrine (PE)-induced cardiac hypertrophy and promoted autophagy activation. Taken together, DJ-1 might repress both pressure overload and PE-induced cardiac hypertrophy via the activation of autophagy.