MilliporeSigma
  • Expression and light-dependent translocation of β-arrestin in the visual system of the terrestrial slug
Expression and light-dependent translocation of β-arrestin in the visual system of the terrestrial slug
The Journal of experimental biology (2017-07-09)
Ryota Matsuo, Yuka Takatori, Shun Hamada, Mitsumasa Koyanagi, Yuko Matsuo
ABSTRACT
Vertebrates, cephalopods and arthropods are equipped with eyes that have the highest spatiotemporal resolution among the animal phyla. In parallel, only animals in these three phyla have visual arrestin specialized for the termination of visual signaling triggered by opsin, in addition to ubiquitously expressed β-arrestin that serves in terminating general G protein-coupled receptor signaling. Indeed, visual arrestin in
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone B-5-1-2, ascites fluid
Sigma-Aldrich
Toluidine Blue, 8.74% (ZN (THEORY))