MilliporeSigma
  • Cardiac striatin interacts with caveolin-3 and calmodulin in a calcium sensitive manner and regulates cardiomyocyte spontaneous contraction rate.
Cardiac striatin interacts with caveolin-3 and calmodulin in a calcium sensitive manner and regulates cardiomyocyte spontaneous contraction rate.
Canadian journal of physiology and pharmacology (2017-08-22)
Moni Nader, Shahd Alotaibi, Ebtehal Alsolme, Bariaa Khalil, Ahmed Abu-Zaid, Rahmah Alsomali, Dana Bakheet, Nduna Dzimiri
ABSTRACT
Impaired cardiomyocyte contraction rate is detrimental to cardiac function and often lethal. Despite advancements in the field, there is a paucity of information regarding the coordination of molecules implicated in regulating the heart rate. Striatin (STRN) is a dynamic protein with binding domains to calmodulin (CaM) and caveolin (Cav), both of which are regulators of myocardial function. However, its role in cardiomyocyte contraction is not yet determined. Herein, we show that STRN is expressed in cardiomyocytes and is more abundant in atrial myocardium than in ventricles. Cardiac expression of STRN (protein and mRNA) was developmentally regulated with the highest expression being at neonatal stage (day one) and the lowest in adult rats (13 weeks). CaM pulldown assay indicated that the interaction of cardiac STRN with CaM and caveolin-3 (Cav-3) was calcium sensitive. Interestingly, the overexpression of STRN induced an increase (∼2-fold) in the rate of the spontaneous contraction of cultured cardiomyocytes, while the knockdown of STRN reduced their contraction rate (∼40%). The expression level of STRN was inversely proportional to the interaction of Cav-3 with the CaM/STRN complex. Collectively, our data delineate a novel role for STRN in regulating cardiomyocyte spontaneous contraction rate and the dynamics of the STRN/Cav-3/CaM complex.
Sigma-Aldrich
RIPA Buffer
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for insect cell culture, suitable for hybridoma
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Calf Serum, from formula-fed bovine calves, iron supplemented, USA origin, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
Tris Buffered Saline, with Tween™ 20, pH 8.0, powder