MilliporeSigma
  • A method for the covalent capture and screening of diverse small molecules in a microarray format.
A method for the covalent capture and screening of diverse small molecules in a microarray format.
Nature protocols (2007-04-05)
James E Bradner, Olivia M McPherson, Angela N Koehler
ABSTRACT
This protocol describes a robust method for the covalent capture of small molecules with diverse reactive functional groups in microarray format, and outlines a procedure for probing small-molecule microarrays (SMMs) with proteins of interest. A vapor-catalyzed, isocyanate-mediated surface immobilization scheme is used to attach bioactive small molecules, natural products and small molecules derived from diversity-oriented synthesis pathways. Additionally, an optimized methodology for screening SMMs with purified proteins and cellular lysates is described. Finally, a suggested model for data analysis that is compatible with commercially available software is provided. These procedures enable a platform capability for discovering novel interactions with potential applications to immunoglobulin profiling, comparative analysis of cellular states and ligand discovery. With the appropriate materials and experimental setup, the printing of SMMs can be completed in 14 hours over 3 days. Screening and data analysis requires 2 days. A detailed timeline is provided.
Sigma-Aldrich
3,5-Dinitrocatechol, solid
Sigma-Aldrich
Monoclonal Anti-Digoxin antibody produced in mouse, clone DI-22, ascites fluid
Sigma-Aldrich
Monoclonal ANTI-FLAG® M5 antibody produced in mouse, clone M5, purified immunoglobulin, buffered aqueous solution
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Sigma-Aldrich
Anti-Corticosterone antibody produced in rabbit, whole antiserum, lyophilized powder